Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 4 trays with 12.5mm polypropylene dividers The trays are 30mm high Each tray holds 420 tubes in 10 x 42 array Each rack holds 1,680 tubes NB U410 holds 3 racks per shelf / 15 racks per freezer NB U570 holds 5 racks per shelf 1 * 1 items
Catalog # AVIAUN132354B/12.5
Supplier:  AVIAMAX
Stainless Steel Rack with 4 trays with 12.5mm polypropylene dividers The trays are 30mm high Each tray holds 420 tubes in 10 x 42 array Each rack holds 1,680 tubes NB U410 holds 3 racks per shelf / 15 racks per freezer NB U570 holds 5 racks per shelf 1 * 1 items
Catalog # AVIAUN132354B/12.5
Supplier:  AVIAMAX
Frequently Bought Together