Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 6 trays Rack holds hinged boxes up to 143w x 156d x 65mm high The trays are 30mm high Each tray holds 4 boxes sideways on, the clip and hinge sit inside the tray Each rack holds 24 boxes NB U360 holds 3 racks per shelf / 9 r 1 * 1 items
  AVIAUXW254106B
 :  AVIAMAX
Stainless Steel Rack with 6 trays Rack holds hinged boxes up to 143w x 156d x 65mm high The trays are 30mm high Each tray holds 4 boxes sideways on, the clip and hinge sit inside the tray Each rack holds 24 boxes NB U360 holds 3 racks per shelf / 9 r 1 * 1 items
  AVIAUXW254106B
 :  AVIAMAX
Frequently Bought Together