Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 4 trays Rack holds hinged boxes up to 143w x 156d x 56mm high Maximum height clearance for boxes is 57mm The trays are 30mm high Each tray holds 3 boxes sideways on, the clip and hinge sit inside the tray Each rack holds 12 1 * 1 items
Catalog # AVIAUXW192354B
Supplier:  AVIAMAX
Stainless Steel Rack with 4 trays Rack holds hinged boxes up to 143w x 156d x 56mm high Maximum height clearance for boxes is 57mm The trays are 30mm high Each tray holds 3 boxes sideways on, the clip and hinge sit inside the tray Each rack holds 12 1 * 1 items
Catalog # AVIAUXW192354B
Supplier:  AVIAMAX
Frequently Bought Together