Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 4 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high Each tray holds 3 boxes Each rack holds 12 boxes 1 * 1 items
Catalog # AVIAUN142354
Supplier:  AVIAMAX
Stainless Steel Rack with 4 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high Each tray holds 3 boxes Each rack holds 12 boxes 1 * 1 items
Catalog # AVIAUN142354
Supplier:  AVIAMAX
Frequently Bought Together