Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 2 trays Rack hold boxes up to 134 x 134 x 115mm The trays are 10mm high Each tray holds 4 boxes Each rack holds 8 boxes NB U570 holds 5 racks per shelf / 25 racks per freezer NB U410 holds 3 racks per shelf / 15 racks per fr 1 * 1 items
Catalog # AVIAUN192352
Supplier:  AVIAMAX
Stainless Steel Rack with 2 trays Rack hold boxes up to 134 x 134 x 115mm The trays are 10mm high Each tray holds 4 boxes Each rack holds 8 boxes NB U570 holds 5 racks per shelf / 25 racks per freezer NB U410 holds 3 racks per shelf / 15 racks per fr 1 * 1 items
Catalog # AVIAUN192352
Supplier:  AVIAMAX
Frequently Bought Together