Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 3 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high Each tray holds 3 boxes Each rack holds 9 boxes 1 * 1 items
Catalog # AVIAUN141773
Supplier:  AVIAMAX
Stainless Steel Rack with 3 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high Each tray holds 3 boxes Each rack holds 9 boxes 1 * 1 items
Catalog # AVIAUN141773
Supplier:  AVIAMAX
Frequently Bought Together