Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 7 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high Each tray holds 4 boxes Each rack holds 28 boxes NB U360 holds 3 racks per shelf / 9 racks per freezer NB U535 holds 4 racks per shelf / 12 racks per fre 1 * 1 items
Catalog # AVIAUN194107
Supplier:  AVIAMAX
Stainless Steel Rack with 7 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high Each tray holds 4 boxes Each rack holds 28 boxes NB U360 holds 3 racks per shelf / 9 racks per freezer NB U535 holds 4 racks per shelf / 12 racks per fre 1 * 1 items
Catalog # AVIAUN194107
Supplier:  AVIAMAX
Frequently Bought Together