Order Entry
United Kingdom
ContactUsLinkComponent
6 components Sodium benzoate [CAS:532-32-1] 200mg/l Saccharin [CAS:81-07-2] 200mg/l Potassium sorbate [CAS:24634615] 200mg/l Acesulfame K [CAS:55589-62-3] 200mg/l Caffeine [CAS:58-08-2] 100mg/l Aspartame [CAS:22839-47-0] 200mg/l in Water : 1 * 500 mL
  LOCA840F.K1.W.L5
 :  Local
6 components Sodium benzoate [CAS:532-32-1] 200mg/l Saccharin [CAS:81-07-2] 200mg/l Potassium sorbate [CAS:24634615] 200mg/l Acesulfame K [CAS:55589-62-3] 200mg/l Caffeine [CAS:58-08-2] 100mg/l Aspartame [CAS:22839-47-0] 200mg/l in Water : 1 * 500 mL
  LOCA840F.K1.W.L5
 :  Local
Frequently Bought Together