Order Entry
United Kingdom
ContactUsLinkComponent
PARD6A (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETE CT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
  ABNOH00050855-PW2
 :  Abnova
PARD6A (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETE CT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
  ABNOH00050855-PW2
 :  Abnova
Frequently Bought Together