Order Entry
United Kingdom
ContactUsLinkComponent
E3 DRYING OVEN,, ,,150 litres capacity ,,Temperature range 40 to 250°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior ,,Easy clean powder coated body ,,Internal dimensions (cms): 52 high x 53 wide x 54 deep ,,External dimensions (cms): 78 h 1 * 1 items
Catalog # GENLE3/OVN/150
Supplier:  GENLAB
E3 DRYING OVEN,, ,,150 litres capacity ,,Temperature range 40 to 250°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior ,,Easy clean powder coated body ,,Internal dimensions (cms): 52 high x 53 wide x 54 deep ,,External dimensions (cms): 78 h 1 * 1 items
Catalog # GENLE3/OVN/150
Supplier:  GENLAB
Frequently Bought Together