Order Entry
United Kingdom
ContactUsLinkComponent
E3 INCUBATOR ,,75 litres capacity ,,Temperature range ambient +5 to 70°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior, easy clean powder coated body ,,Internal dimensions (cms): 46 high x 49 wide x 33 deep ,,External dimensions (cms): 72 h 1 * 1 items
Catalog # GENLE3/1NC/75
Supplier:  GENLAB
E3 INCUBATOR ,,75 litres capacity ,,Temperature range ambient +5 to 70°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior, easy clean powder coated body ,,Internal dimensions (cms): 46 high x 49 wide x 33 deep ,,External dimensions (cms): 72 h 1 * 1 items
Catalog # GENLE3/1NC/75
Supplier:  GENLAB
Frequently Bought Together