Pyrotell-T Multi-Test Turbidimetric Formulation, 25 vials x 5ml/vial (approx 1250 tests) 1 * 5 items
Catalog # CAPET0051-25
Supplier: Associates of Cape Cod
Pyrotell-T Multi-Test Turbidimetric Formulation, 25 vials x 5ml/vial (approx 1250 tests) 1 * 5 items
Catalog # CAPET0051-25
Supplier: Associates of Cape Cod
Frequently Bought Together
High-sensitivity coliform count plates, Petrifilm™
Supplier: 3M Food Safety
Control Standard Endotoxin (CSE) E.coli O113:H10 1 * 0,5 µG
Supplier: Associates of Cape Cod
YEAST NITROGEN BASE, FOR MICROBIOLOGY* 1 * 100 g
Supplier: Merck
TCBS AGAR, FOR MICROBIOLOGY 1 * 500 g
Supplier: Merck
DNA Loading Dye
Supplier: STEMCELL Technologies
Culti-Loops™ <i>Moraxella catarrhalis</i> ATCC™ 8176™
Supplier: THERMO FISHER DIAGNOSTICS
Culti-Loops™ <i>Tatlockia micdadei</i> ATCC™ 33204™
Supplier: THERMO FISHER DIAGNOSTICS
Annexin V Apoptosis Detection Kit with 7-AAD, FITC
Supplier: STEMCELL Technologies
VWR® Contact Plates, Blister Packed, Irradiated
Supplier: VWR Chemicals
Coagulase plasma
Supplier: THERMO FISHER DIAGNOSTICS
Culture media, ready to use, in plates
Supplier: THERMO FISHER DIAGNOSTICS
Keyboard for medical environments, InduProof® Smart Classic
Supplier: GETT GERATETECHNIK