Order Entry
United Kingdom
ContactUsLinkComponent
PPARG Human Recombinant encoding amino acids 1-477 expressed in E.coli, is fused to GST tag and it is antibody reactive.The PPARG is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80861-2
 :  Biorbyt
PPARG Human Recombinant encoding amino acids 1-477 expressed in E.coli, is fused to GST tag and it is antibody reactive.The PPARG is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80861-2
 :  Biorbyt
Frequently Bought Together