Dispersive 2ml,universal kit w/ch,1/pk 5mgpsa, 5mgc18, 7.5mggcb, 15mgmgso4 1 * 100 items
HEWL5982-0028CH
: Agilent
Dispersive 2ml,universal kit w/ch,1/pk 5mgpsa, 5mgc18, 7.5mggcb, 15mgmgso4 1 * 100 items
HEWL5982-0028CH
: Agilent
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection