Order Entry
United Kingdom
ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-03M15K
C18G x 3µm particle size x 120Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-03M15K
Frequently Bought Together