Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Okadaic acid

Okadaic acid

100 µG
Catalog # CAYM10011490-100
Supplier:   Cayman Chemical
Okadaic acid
Catalog # CAYM10011490-100
Supplier:  Cayman Chemical
CAS Number:  78111-17-8
Okadaic acid

100 µG

CAS Number:  78111-17-8
Price
£388.00
Log in for availability
1 * 100 µG
Product Details & Documents
Synonyms:  9,10-deepithio-9,10-didehydroacanthifolicin, OA
  • CAS Number:78111-17-8
  • Formula:C₄₄H₆₈O₁₃
  • MDL Number:MFCD00083455
  • Molecular Weight:805.02 g/mol
Specifications

Specifications

  • Pk:
    100 µG

Specifications