Okadaic acid
50 µG
Catalog # CAYM10011490-50
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
- CAS Number:78111-17-8
- Formula:C₄₄H₆₈O₁₃
- MDL Number:MFCD00083455
- Molecular Weight:805.02 g/mol
Specifications
Specifications
- Pk:50 µG
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



