Order Entry
United Kingdom
ContactUsLinkComponent
Slimline Single wavelength slide-out transilluminator (20 x 20cm 365nm) 230V, 50Hz. Longwave - with runners (no cover) 6 tubes. For use with IG and UG systems 1 * 1 items
Catalog # SYNOGE-2020L-E
Slimline Single wavelength slide-out transilluminator (20 x 20cm 365nm) 230V, 50Hz. Longwave - with runners (no cover) 6 tubes. For use with IG and UG systems 1 * 1 items
Catalog # SYNOGE-2020L-E
Frequently Bought Together