Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Scissors No. 145, Feather®
Scissors No. 145, Feather®
Sharp/sharp
Catalog # FEAT208000157
Supplier:   PFM Medical GmbH
Scissors No. 145, Feather®
Catalog # FEAT208000157
Supplier:  PFM Medical GmbH
Scissors No. 145, Feather®
Scissors No. 145, Feather®
Sharp/sharp
Price
£123.00
Log in for availability
1 * 5 PAIR
Product Details & Documents
Pathology scissors made of stainless steel with easily exchangeable blades.
  • No regrinding
  • Easy blade change
Documents
Specifications
  • Type:
    Sharp/sharp