PATHHUNTER CELL PLATING 5 REAGENT 1 * 100 mL
Catalog # DISX93-0563R5A
Supplier: DISCOVERX
Frequently Bought Together
VWR®, Igientest HACCP
Supplier: VWR Collection
Accessories for Anaerobic Systems, Oxoid
Supplier: THERMO FISHER DIAGNOSTICS
Culti-Loops™ <i>Myroides odoratus</i> ATCC™ 4651™
Supplier: THERMO FISHER DIAGNOSTICS
SWABS WITH BPW BUFFER 1 * 500 items
Supplier: Shimadzu Diagnostics Europe
Contact plates, RT/RTplus, single bagged
Supplier: MERCK LIFE SCIENCE
Prepared media , bottled culture media
Supplier: THERMO FISHER DIAGNOSTICS
VWR® Culture Media, Ready to Use, in Bottles
Supplier: VWR Chemicals
Reinforced Clostridial Agar 1 * 500 g
Supplier: Merck
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
GIFU ANAEROBIC BROTH MODIFIED (GAM) 1 * 500 g
Supplier: HIMEDIA
Agar
Supplier: Roth Carl
E.coli OD10 (10x Concentrate for preparation of Rich Growth Media) 13C 1 * 1 L
Supplier: MARKET SOURCE PART PROCESS
YM AGAR SLANT TUBES 1 * 20 items
Supplier: VWR Chemicals
SABOURAUD DEXTROSE AGAR 1 * 10 items
Supplier: THERMO FISHER DIAGNOSTICS