5-Bromo-3-hydroxyisoquinoline
Bottle, 500 mg
Catalog # APOSOR300065-500MG
Supplier: Apollo Scientific
5-Bromo-3-hydroxyisoquinoline
Catalog # APOSOR300065-500MG
Supplier: Apollo Scientific
CAS Number: 1175271-86-9
Jump to:
Product Details & Documents
Synonyms: 5-Bromoisoquinolin-3-ol, 5-Bromo-3-hydroxy-2-azanaphthalene
- CAS Number:1175271-86-9
- Formula:C₉H₆BrNO
- MDL Number:MFCD11040413
- Molecular Weight:224.054 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:500 mg



