Sterilisation wrap
Sterilisation wrap, crepe paper, 60 gsm, White, 130×150 mm
Supplier: DISTRIBUTOR
Jump to:
Product Details & Documents
Sterilisation wrap is available in embossed crepe or plain papers.
: Manufactured under BS EN ISO.9001.2008 and BS EN ISO 13485:2003 and conforms to BS EN ISO11607-1:2006 part 1 and BS EN 868:2009 part 2.
Specifications
- Colour:White
- Description:Sterilisation wrap, crepe paper, 60 gsm
- Dimensions:130×150 mm
- Seq:01