Methylated Spirits 25L 98/99% W/W (Clear) (authorisation required) 74OP 1 * 25 L
Catalog # DIUKME38000-355
Supplier: DISTRIBUTOR
Methylated Spirits 25L 98/99% W/W (Clear) (authorisation required) 74OP 1 * 25 L
Catalog # DIUKME38000-355
Supplier: DISTRIBUTOR
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Diisopropyl ether, Sigma-Aldrich®
Supplier: Merck
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Methyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Nitromethane 96% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethylene glycol, Sigma-Aldrich®
Supplier: Merck
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific