Order Entry
United Kingdom
ContactUsLinkComponent
Gas50co,2.5%ch4,18%o2,n2 110l 3g18x25w50m-n2110 1 * 1 items
Catalog # BWTE3G18X25W50M-N2
Supplier:  BW TECHNOLOGIES
Gas50co,2.5%ch4,18%o2,n2 110l 3g18x25w50m-n2110 1 * 1 items
Catalog # BWTE3G18X25W50M-N2
Supplier:  BW TECHNOLOGIES
Frequently Bought Together