Order Entry
United Kingdom
ContactUsLinkComponent
WBI-2010 WITH AFC ***221-73041-48*** Wide Bore Injektor with integrated AFC-H (High Pressure Advanced Flow Control) for electonic control of all carrier gas parameters (please order column hanger p/n 221-47159 separately) 1 * 1 items
Catalog # SHIM221-71041-38
Supplier:  SHIMADZU
WBI-2010 WITH AFC ***221-73041-48*** Wide Bore Injektor with integrated AFC-H (High Pressure Advanced Flow Control) for electonic control of all carrier gas parameters (please order column hanger p/n 221-47159 separately) 1 * 1 items
Catalog # SHIM221-71041-38
Supplier:  SHIMADZU
Frequently Bought Together