Trypsin / EDTA (0.04% / 0.03%) 1 * 125 mL
Catalog # C-41010
Supplier: PROMOCELL
Frequently Bought Together
Counting chambers, C-Chip
Supplier: NANOENTEK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Sampling tube (betw.needle / injec valve 1 * 1 items
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Caps for microcentrifuge tubes with thread
Supplier: Simport Scientific
96-well solid black and white polystyrene microplates, Corning®
Supplier: Corning
Container with screw cap
Supplier: SARSTEDT
Sampling scoops, LaboPlast®
Supplier: BURKLE
HEPES Buffered Saline Solution 1 * 250 mL
Supplier: PROMOCELL
Lids for Nunc™ microplates
Supplier: Thermo Fisher Scientific
ViewPlate-384 blk,clr btm,Tissue Culture 1 * 40 items
Supplier: PerkinElmer
Trypsin neutralizing solution
Supplier: PROMOCELL
Test tubes, disposable
Supplier: SEMADENI
Cryo.S™ tubes, 1 and 2 ml
Supplier: Greiner Bio-One