Millesimal buffer solutions
10,01, ±0,02, 25 °C, Sachet, 25×20 ml, 25
HANNHI60010-02
: Hanna
These millesimal buffer solution are NIST traceable.
- Colour coded label for easy pH buffer value identification
- Temperature chart of the actual pH value at various temperatures printed on each sachet
Accordance with ISO 3696/BS3978 standards using high purity salts, deionised water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers.
Delivery information: The buffers in sachets are delivered with a certificate of conformity.
- Pack type:Sachet
- Pk:25
- Reference temperature:25 °C
- pH accuracy:±0,02
- Pk Info:25×20 ml
- pH value:10,01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



