Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Millesimal buffer solutions

Millesimal buffer solutions

1,67, ±0,02, 25 °C, Sachet, 25×20 ml, 25
Catalog # HANNHI60016-02
Supplier:   Hanna
Millesimal buffer solutions
Catalog # HANNHI60016-02
Supplier:  Hanna
CAS Number:  
Millesimal buffer solutions

1,67, ±0,02, 25 °C, Sachet, 25×20 ml, 25

CAS Number:  
Price
£135.00
Log in for availability
1 * 25 items
Product Details & Documents

These millesimal buffer solution are NIST traceable.

  • Colour coded label for easy pH buffer value identification
  • Temperature chart of the actual pH value at various temperatures printed on each sachet

Accordance with ISO 3696/BS3978 standards using high purity salts, deionised water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers.

Delivery information: The buffers in sachets are delivered with a certificate of conformity.

Specifications

Specifications

  • Pack type:
    Sachet
  • Pk:
    25
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,02
  • Pk Info:
    25×20 ml
  • pH value:
    1,67

Specifications