RAT/MOUSE INSULIN ELISA 1 * 1 items
Catalog # EZRMI-13K
Supplier: Merck Millipore (Calbiochem)
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Sampling Scoops
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
96 Well Cluster Tubes, Corning®
Supplier: Corning
Roller bottle standard, surface treated, 1000 ml vent cap 1 * 24 items
Supplier: GUANGZHOU JET BIOFILTRATION
Transport swabs (Sterilin)
Supplier: STERILIN
Vial centrifuge ster pp 50ml 30x115mm 1 * 25 items
Supplier: SEMADENI
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Aspirating pipettes, Falcon®
Supplier: Corning
Polypropylene centrifuge tubes, PerformR®
Supplier: LABCON
Racks for Upright Freezers with Drawers
Supplier: GLW
Test tubes
Supplier: KARTELL
Closures for test tubes and flasks
Supplier: JAECE INDUSTRIES
CELLSTAR PLATE CELL REPELLENT 96W 39 1 * 32 items
Supplier: Greiner Bio-One
Microplates, 384-well
Supplier: Greiner Bio-One
Inserts for Brand® cryogenic tubes
Supplier: Brand