3'-Fluoro-4'-hydroxyacetophenone
Glass bottle for solids, 25 g
Catalog # ACTIAS23052G25
Marketsource
Supplier: ACTIVATE SCIENTIFIC
3'-Fluoro-4'-hydroxyacetophenone
Catalog # ACTIAS23052G25
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 403-14-5
Jump to:
Product Details & Documents
Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone
- CAS Number:403-14-5
- Formula:C₈H₇FO₂
- MDL Number:MFCD00215840
- Molecular Weight:154.14 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



