3'-Fluoro-4'-hydroxyacetophenone
Glass bottle for solids, 100 g
Catalog # ACTIAS23052G100
Marketsource
Supplier: ACTIVATE SCIENTIFIC
3'-Fluoro-4'-hydroxyacetophenone
Catalog # ACTIAS23052G100
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 403-14-5
Jump to:
Product Details & Documents
Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone
- CAS Number:403-14-5
- Formula:C₈H₇FO₂
- MDL Number:MFCD00215840
- Molecular Weight:154.14 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



