3-(4-Cyanophenoxy)propionic acid
Glass bottle for solids, 5 g
Catalog # ACTIAS22929G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
3-(4-Cyanophenoxy)propionic acid
Catalog # ACTIAS22929G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 58228-89-0
Jump to:
Product Details & Documents
- CAS Number:58228-89-0
- Formula:C₁₀H₉NO₃
- MDL Number:MFCD09043034
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



