3-(4-Cyanophenoxy)propionic acid
Glass bottle for solids, 1 g
Catalog # ACTIAS22929G1
Marketsource
Supplier: ACTIVATE SCIENTIFIC
3-(4-Cyanophenoxy)propionic acid
Catalog # ACTIAS22929G1
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 58228-89-0
Jump to:
Product Details & Documents
- CAS Number:58228-89-0
- Formula:C₁₀H₉NO₃
- MDL Number:MFCD09043034
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



