6-Nitroindole
Glass bottle for solids, 5 g
Catalog # ACTIAS21033G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
6-Nitroindole
Catalog # ACTIAS21033G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 4769-96-4
Jump to:
Product Details & Documents
Synonyms: 6-Nitro-1H-indole
- CAS Number:4769-96-4
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00051497
- Molecular Weight:162.15 g/mol
- EINECS:225-311-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Cat. No.:ACTIAS21033G5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



