6-Nitroindole
Glass bottle for solids, 25 g
Catalog # ACTIAS21033G25
Marketsource
Supplier: ACTIVATE SCIENTIFIC
6-Nitroindole
Catalog # ACTIAS21033G25
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 4769-96-4
Jump to:
Product Details & Documents
Synonyms: 6-Nitro-1H-indole
- CAS Number:4769-96-4
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00051497
- Molecular Weight:162.15 g/mol
- EINECS:225-311-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



