Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Bromothiazole
Glass bottle for solids, 100 g
  ACTIAS50234G100
Marketsource
2-Bromothiazole
  ACTIAS50234G100
Marketsource
 :  3034-53-5
2-Bromothiazole
Glass bottle for solids, 100 g
 :  3034-53-5
1 * 100 g
  •  :3034-53-5
  •  :C₃H₂BrNS
  •  :MFCD00005316
  •  :164.03 g/mol
  •  :171 °C (1013 hPa)
  •  :1,82 g/cm³ (20 °C)
  •  :221-229-4
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g