2-(4-Fluorophenyl)ethanol
Glass bottle for solids, 25 g
Catalog # ACTIAS24775G25
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-(4-Fluorophenyl)ethanol
Catalog # ACTIAS24775G25
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 7589-27-7
Jump to:
Product Details & Documents
Synonyms: 4-Fluorophenethyl alcohol
- CAS Number:7589-27-7
- Formula:C₈H₉FO
- MDL Number:MFCD00002898
- Molecular Weight:140.16 g/mol
- Boiling Pt:59…61 °C (0,5 mmHg)
- Boiling Pt:59…61 °C (0.5 mmHg)
- Density:1,137 g/cm³ (20 °C)
- EINECS:231-499-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



