Order Entry
United Kingdom
ContactUsLinkComponent
2,4-DICHLORO-5-METHYLTHIENO[2,3-D]PYRIMIDINE MFCD09965519 PURITY: 95+% MOLECULAR FORMULA C7H4CL2N2S MOLECULAR WEIGHT 219.09 CAS: 56844-38-3 1 * 1 g
Catalog # ACTIAS2584G1
2,4-DICHLORO-5-METHYLTHIENO[2,3-D]PYRIMIDINE MFCD09965519 PURITY: 95+% MOLECULAR FORMULA C7H4CL2N2S MOLECULAR WEIGHT 219.09 CAS: 56844-38-3 1 * 1 g
Catalog # ACTIAS2584G1
Frequently Bought Together