2,4-DICHLORO-6-(TRIFLUOROMETHYL)QUINAZOLINE MFCD09954876 PURITY: 95+% C9H3Cl2F3N2 1 * 1 g
Catalog # ACTIAS2568G1
Supplier: ACTIVATE SCIENTIFIC
2,4-DICHLORO-6-(TRIFLUOROMETHYL)QUINAZOLINE MFCD09954876 PURITY: 95+% C9H3Cl2F3N2 1 * 1 g
Catalog # ACTIAS2568G1
Supplier: ACTIVATE SCIENTIFIC
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK