Order Entry
United Kingdom
ContactUsLinkComponent
Manual Tissue Arrayer MTA-1 Specifications: Speed: cores/hour: 30-70 Max block capacity: 4(with indexer - optional) Smallest recommended punch size: 0.6 Donor block height: 3 cm 1 * 1 items
Catalog # LOCAGX-BI-MTA-1
Supplier:  Local
Manual Tissue Arrayer MTA-1 Specifications: Speed: cores/hour: 30-70 Max block capacity: 4(with indexer - optional) Smallest recommended punch size: 0.6 Donor block height: 3 cm 1 * 1 items
Catalog # LOCAGX-BI-MTA-1
Supplier:  Local
Frequently Bought Together