RNA sample loading buffer, no ethidium bromide
RNA Sample Loading Buffer, no Ethidium Bromide, 1 mL
Supplier: Thermo Fisher Scientific
RNA sample loading buffer, no ethidium bromide
Catalog # J61937.XF
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00213041
Applications: Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide
Specifications
Specifications
- Pk:1 mL
- Description:RNA Sample Loading Buffer, no Ethidium Bromide
- Cat. No.:J61937.XF
- Physical form:Liquid
- MDL No.:MFCD00213041



