Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Buffers, MOPS, 0,5 M solutions

Buffers, MOPS, 0,5 M solutions

MOPS buffer solution, 0,5 mol/L, 7,5, Glass bottle, 23, 250 mL
Catalog # J62839.AK
Buffers, MOPS, 0,5 M solutions
Catalog # J62839.AK
CAS Number:  
Buffers, MOPS, 0,5 M solutions
Buffers, MOPS, 0,5 M solutions

MOPS buffer solution, 0,5 mol/L, 7,5, Glass bottle, 23, 250 mL

CAS Number:  
Price
£90.80
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    0,5 mol/L
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    MOPS buffer solution
  • Pk Info:
    23
  • pH value:
    7,5
  • Cat. No.:
    J62839.AK
  • Odour/scent:
    Odourless
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006183
  • CAS No.:
    1132-61-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.