Order Entry
United Kingdom
ContactUsLinkComponent
HistoTainer 120 mL, Non Tampered Evident - Bulk, Prefilled, Specimen Containers. Made of virgin, translucent polypropylene / Closure made of polyethylene, designed for collection, transport and storage of histology specimens. Closures are manufactur 1 * 96 items
Catalog # SIMPM959-120FMA
Supplier:  Simport Scientific
HistoTainer 120 mL, Non Tampered Evident - Bulk, Prefilled, Specimen Containers. Made of virgin, translucent polypropylene / Closure made of polyethylene, designed for collection, transport and storage of histology specimens. Closures are manufactur 1 * 96 items
Catalog # SIMPM959-120FMA
Supplier:  Simport Scientific
Frequently Bought Together