Order Entry
United Kingdom
ContactUsLinkComponent
Dimensions,,, ,,,,,,92x56x46mm ErP Compliance, ,,,,Non Compliant Input Voltage, ,,,,,,230V ac Medical Approval, ,,,,No Number of Outputs, ,,,,1 Operating Temperature, ,,0 +25C Output Current, ,,,,1000 1000 800 800 700 600 500mA Output Voltage 1 * 1 items
Catalog # RSCO615-8919
Supplier:  RS COMPONENTS
Dimensions,,, ,,,,,,92x56x46mm ErP Compliance, ,,,,Non Compliant Input Voltage, ,,,,,,230V ac Medical Approval, ,,,,No Number of Outputs, ,,,,1 Operating Temperature, ,,0 +25C Output Current, ,,,,1000 1000 800 800 700 600 500mA Output Voltage 1 * 1 items
Catalog # RSCO615-8919
Supplier:  RS COMPONENTS
Frequently Bought Together