Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Temperature sensors

Thermocouple rod, type J (Fe-CuNi) and cable (1,2 m), +500…+1000 °C, 1,6×300 mm
Catalog # WINKWFSJ1500-XX012
Supplier:   WINKLER
Temperature sensors
Catalog # WINKWFSJ1500-XX012
Supplier:  WINKLER
Temperature sensors
Temperature sensors

Thermocouple rod, type J (Fe-CuNi) and cable (1,2 m), +500…+1000 °C, 1,6×300 mm

Price
Inquire For Price
Log in for availability
Product Details & Documents
Specifications
  • Range:
    +500…+1000 °C
  • Description:
    Thermocouple rod, type J (Fe-CuNi) and cable (1,2 m)
  • Seq:
    05
  • Ø×L:
    1,6×300 mm