Conductivity meter Portavo 904X digital with analog sensor input and digital Memosens input, sensor quiver, protective hood, clear glass display, waterproof IP66/67, data logger 5.000 values, Micro USB for PC connection, Paraly 112 PC software, ATEX 1 * 1 items
Catalog # 663-0226
Supplier: KNICK