Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Oleic acid

Glass bottle for solids, 10 g
Catalog # CAYM90260-10
Supplier:   Cayman Chemical
Oleic acid
Catalog # CAYM90260-10
Supplier:  Cayman Chemical
CAS Number:  112-80-1
Oleic acid
Oleic acid

Glass bottle for solids, 10 g

CAS Number:  112-80-1
Price
£199.00
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Merck Index:13,06898
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:16.7 °C
  • Density:0.8946 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Storage Temperature:Freezer
  • EINECS:204-007-1

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications