Conductivity standards
Conductivity standard, 84 uS, 500 mL
JENW16156-80
Standard solutions are used for calibrating conductivity cells.
- Pk:500 mL
- Description:Conductivity standard
- Conductivity:84 uS
Standard solutions are used for calibrating conductivity cells.