2,7-Dinitro-9-fluorenone ≥97%
Glass bottle for solids, 5 g
Catalog # H55176.06
Supplier: Thermo Fisher Scientific
2,7-Dinitro-9-fluorenone ≥97%
Catalog # H55176.06
Supplier: Thermo Fisher Scientific
CAS Number: 31551-45-8
Jump to:
Product Details & Documents
- CAS Number:31551-45-8
- Formula:C₁₃H₆N₂O₅
- MDL Number:MFCD00001153
- Molecular Weight:270.2 g/mol
- Melting Pt:292…295 °C
- Storage Temperature:Ambient
- EINECS:250-695-1
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



