Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Printer and balance accessories
Printer and balance accessories
Bubble level for ranger
Catalog # 611-3748
Supplier:   OHAUS
Printer and balance accessories
Catalog # 611-3748
Supplier:  OHAUS
Printer and balance accessories
Printer and balance accessories
Bubble level for ranger
Price
£11.70
Log in for availability
1 * 1 items
Product Details & Documents
The SF40A impact printer and the STP103 thermal printer are compatible with any Ohaus balance with an RS232 interface. They are particularly useful when coupled to an Ohaus balance with GLP functionality as they can provide a hard copy audit trail. A printer cable is required (but not supplied) for specific Ohaus balances.
Ordering information: There are a wide variety of accessories and replacement parts for Ohaus balances and the most commonly used are listed below: For further items please visit the local Avantor website.
Specifications
  • Description:
    Bubble level for ranger
  • Seq:
    14